Assay Details
Amplicon Length: | 57 |
Chromosome Location: | Chr. 22 - 46112947 - 46112947 [-] on 46112947 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-let-7a-3p | MIMAT0004481 |
| Human | hsa-let-7a-5p | MIMAT0000062 |
Stem-loop Details
Human

Genomic Map