Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. X - 147237418 - 147237418 [+] on 147237418 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-508-5p | MIMAT0004778 |
| Human | hsa-miR-508-3p | MIMAT0002880 |
Stem-loop Details
Human

Genomic Map