Assay Details
Amplicon Length: | 83 |
Chromosome Location: | Chr. 5 - 169263897 - 169263897 [+] on 169263897 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-585-5p | MIMAT0026618 |
Human | hsa-miR-585-3p | MIMAT0003250 |
Stem-loop Details
Human
