Assay Details
Amplicon Length: | 76 |
Chromosome Location: | Chr. 10 - 96829171 - 96829171 [+] on 96829171 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-607 | MIMAT0003275 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 76 |
Chromosome Location: | Chr. 10 - 96829171 - 96829171 [+] on 96829171 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-607 | MIMAT0003275 |