Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. 15 - 70079772 - 70079772 [+] on 70079772 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-629-5p | MIMAT0004810 |
| Human | hsa-miR-629-3p | MIMAT0003298 |
Stem-loop Details
Human

Genomic Map