Assay Details
Amplicon Length: | 88 |
Chromosome Location: | Chr. 22 - 37847469 - 37847469 [+] on 37847469 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-659-3p | MIMAT0003337 |
| Human | hsa-miR-659-5p | MIMAT0022710 |
Stem-loop Details
Human

Genomic Map