Assay Details
Amplicon Length: | 82 |
Chromosome Location: | Chr. 2 - 25328447 - 25328447 [+] on 25328447 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1301-5p | MIMAT0026639 |
| Human | hsa-miR-1301-3p | MIMAT0005797 |
Stem-loop Details
Human

Genomic Map