Assay Details
Amplicon Length: | 69 |
Chromosome Location: | Chr. 5 - 110513532 - 110513532 [+] on 110513532 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548f-3p | MIMAT0005895 |
Stem-loop Details
Human

Genomic Map