Assay Details
Amplicon Length: | 66 |
Chromosome Location: | Chr. 6 - 138435065 - 138435065 [+] on 138435065 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3145-5p | MIMAT0019205 |
| Human | hsa-miR-3145-3p | MIMAT0015016 |
Stem-loop Details
Human

Genomic Map