Assay Details
Amplicon Length: | 94 |
Chromosome Location: | Chr. 16 - 81385259 - 81385259 [-] on 81385259 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4720-5p | MIMAT0019833 |
| Human | hsa-miR-4720-3p | MIMAT0019834 |
Stem-loop Details
Human

Genomic Map