Assay Details
Amplicon Length: | 189 |
Chromosome Location: | Chr. 2 - 35471059 - 35471059 [+] on 35471059 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548ad-3p | MIMAT0018946 |
| Human | hsa-miR-548ad-5p | MIMAT0032114 |
Stem-loop Details
Human

Genomic Map