Assay Details
Amplicon Length: | 176 |
Chromosome Location: | Chr. 2 - 184378819 - 184378819 [-] on 184378819 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548ae-3p | MIMAT0018954 |
Stem-loop Details
Human

Genomic Map