Assay Details
Amplicon Length: | 86 |
Chromosome Location: | Chr. 5 - 58529595 - 58529595 [-] on 58529595 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548ae-3p | MIMAT0018954 |
Human | hsa-miR-548ae-5p | MIMAT0032115 |
Stem-loop Details
Human
