Assay Details
Amplicon Length: | 118 |
Chromosome Location: | Chr. 4 - 76575301 - 76575301 [+] on 76575301 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548ah-5p | MIMAT0018972 |
| Human | hsa-miR-548ah-3p | MIMAT0020957 |
Stem-loop Details
Human

Genomic Map