Assay Details
Amplicon Length: | 108 |
Chromosome Location: | Chr. 6 - 132115043 - 132115043 [+] on 132115043 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548aj-3p | MIMAT0018990 |
Stem-loop Details
Human

Genomic Map