Assay Details
Amplicon Length: | 59 |
Chromosome Location: | Chr. 9 - 36864503 - 36864503 [-] on 36864503 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4540 | MIMAT0019083 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 59 |
Chromosome Location: | Chr. 9 - 36864503 - 36864503 [-] on 36864503 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-4540 | MIMAT0019083 |