Assay Details
Amplicon Length: | 77 |
Chromosome Location: | Chr. X - 16626862 - 16626862 [+] on 16626862 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548am-5p | MIMAT0022740 |
| Human | hsa-miR-548am-3p | MIMAT0019076 |
Stem-loop Details
Human

Genomic Map