Assay Details
Amplicon Length: | 78 |
Chromosome Location: | Chr. 13 - 65966582 - 65966582 [+] on 65966582 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548x-3p | MIMAT0015081 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 78 |
Chromosome Location: | Chr. 13 - 65966582 - 65966582 [+] on 65966582 Build GRCh38 |
Species: | Human |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548x-3p | MIMAT0015081 |