Assay Details
Amplicon Length: | 60 |
Chromosome Location: | Chr. 18 - 72853142 - 72853142 [+] on 72853142 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-548av-5p | MIMAT0022303 |
| Human | hsa-miR-548av-3p | MIMAT0022304 |
Stem-loop Details
Human

Genomic Map