Assay Details
Gene Family ID: | MIPF0000069, mir-32 |
Mature miRNA Sequence: | UAUUGCACAUUACUAAGUUGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Callithrix jacchus | cja-miR-32 | MIMAT0039452 |
| Chicken | gga-miR-32-5p | MIMAT0001125 |
| Cricetulus griseus | cgr-miR-32-5p | MIMAT0023904 |
| Gorilla gorilla | ggo-miR-32 | MIMAT0002387 |
| Macaca nemestrina | mne-miR-32 | MIMAT0002390 |
| Monodelphis domestica | mdo-miR-32-5p | MIMAT0004095 |
| View More Rows | ||
| Pan paniscus | ppa-miR-32 | MIMAT0002391 |
| Pan troglodytes | ptr-miR-32 | MIMAT0002386 |
| Pig | ssc-miR-32 | MIMAT0002149 |
| Pongo pygmaeus | ppy-miR-32 | MIMAT0002388 |
| Pteropus alecto | pal-miR-32-5p | MIMAT0040357 |
| Python bivittatus | pbv-miR-32-5p | MIMAT0038881 |
| Rhesus monkey | mml-miR-32-5p | MIMAT0002385 |
| Saguinus labiatus | sla-miR-32 | MIMAT0002389 |
| Taeniopygia guttata | tgu-miR-32 | MIMAT0014584 |
Stem-loop Details
Callithrix jacchus
Chicken
Cricetulus griseus
Gorilla gorilla
Macaca nemestrina
Monodelphis domestica
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Python bivittatus
Rhesus monkey
Saguinus labiatus
Taeniopygia guttata
More Information
Related Products
miRBase Alias:
Gallus gallus: gga-miR-32 (v19)
Macaca mulatta: mml-miR-32 (v19)
Monodelphis domestica: mdo-miR-32 (v19)
Macaca mulatta: mml-miR-32 (v19)
Monodelphis domestica: mdo-miR-32 (v19)

Genomic Map