Assay Details
Gene Family ID: | MIPF0000073, mir-129 |
Mature miRNA Sequence: | CUUUUUGCGGUCUGGGCUUGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-miR-129 | MIMAT0009220 |
| Bovine | bta-miR-129-5p | MIMAT0009221 |
| Eptesicus fuscus | efu-miR-129b | MIMAT0035105 |
| Fugu rubripes | fru-miR-129 | MIMAT0003035 |
| Hippoglossus hippoglossus | hhi-miR-129a | MIMAT0030351 |
| Hippoglossus hippoglossus | hhi-miR-129b-5p | MIMAT0030349 |
| View More Rows | ||
| Monodelphis domestica | mdo-miR-129-5p | MIMAT0004105 |
| Ophiophagus hannah | oha-miR-129a-5p | MIMAT0036680 |
| Tetraodon nigroviridis | tni-miR-129 | MIMAT0003036 |
| Zebrafish | dre-miR-129-5p | MIMAT0001825 |
Stem-loop Details
Bovine
Eptesicus fuscus
Fugu rubripes
Hippoglossus hippoglossus
Monodelphis domestica
Ophiophagus hannah
Tetraodon nigroviridis
Zebrafish
More Information
Related Products
miRBase Alias:
Danio rerio: dre-miR-129 (v18)
Monodelphis domestica: mdo-miR-129 (v19)
Monodelphis domestica: mdo-miR-129 (v19)

Genomic Map