Assay Details
Gene Family ID: | MIPF0000095, mir-122 |
Mature miRNA Sequence: | UGGAGUGUGACAAUGGUGUUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-122-5p | MIMAT0000421 |
| Mouse | mmu-miR-122-5p | MIMAT0000246 |
| Rat | rno-miR-122-5p | MIMAT0000827 |
| Alligator mississippiensis | ami-miR-122-5p | MIMAT0038123 |
| Anolis carolinensis | aca-miR-122-5p | MIMAT0021723 |
| Astatotilapia burtoni | abu-miR-122 | MIMAT0042117 |
| View More Rows | ||
| Bovine | bta-miR-122 | MIMAT0003849 |
| Callithrix jacchus | cja-miR-122 | MIMAT0039630 |
| Chrysemys picta | cpi-miR-122-5p | MIMAT0037724 |
| Columba livia | cli-miR-122-5p | MIMAT0038491 |
| Dasypus novemcinctus | dno-miR-122-5p | MIMAT0047610 |
| Dog | cfa-miR-122 | MIMAT0006619 |
| Fugu rubripes | fru-miR-122 | MIMAT0002991 |
| Gadus morhua | gmo-miR-122-5p | MIMAT0044063 |
| Goat | chi-miR-122 | MIMAT0035918 |
| Gorilla gorilla | ggo-miR-122 | MIMAT0024179 |
| Guinea pig | cpo-miR-122-5p | MIMAT0046961 |
| Horse | eca-miR-122 | MIMAT0012995 |
| Metriaclima zebra | mze-miR-122 | MIMAT0042349 |
| Neolamprologus brichardi | nbr-miR-122 | MIMAT0042551 |
| Oreochromis niloticus | oni-miR-122 | MIMAT0042727 |
| Ornithorhynchus anatinus | oan-miR-122-5p | MIMAT0006825 |
| Pan troglodytes | ptr-miR-122 | MIMAT0007963 |
| Paralichthys olivaceus | pol-miR-122-5p | MIMAT0025428 |
| Pongo pygmaeus | ppy-miR-122 | MIMAT0015746 |
| Pteropus alecto | pal-miR-122-5p | MIMAT0039776 |
| Pundamilia nyererei | pny-miR-122 | MIMAT0042894 |
| Python bivittatus | pbv-miR-122-5p | MIMAT0038909 |
| Rabbit | ocu-miR-122-5p | MIMAT0048201 |
| Rhesus monkey | mml-miR-122a-5p | MIMAT0006180 |
| Salmo salar | ssa-miR-122-5p | MIMAT0032301 |
| Taeniopygia guttata | tgu-miR-122-5p | MIMAT0014529 |
| Tetraodon nigroviridis | tni-miR-122 | MIMAT0002992 |
| Zebrafish | dre-miR-122 | MIMAT0001818 |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-122 |
| Stem-loop Accession # | MI0000442 |
| Stem-loop Sequence | CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGGC |
| Chromosome Location | Chr. 18 - 58451074 - 58451158 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000421
miRBase ID: hsa-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: Chr. 18 - 58451074 - 58451158 [+] on Build GRCh38 |
| Stem-loop ID | mmu-mir-122 |
| Stem-loop Accession # | MI0000256 |
| Stem-loop Sequence | AGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAACCAUCAAACGCCAUUAUCACACUAAAUAGCU |
| Chromosome Location | Chr. 18 - 65248861 - 65248926 [+] on Build GRCm38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000246
miRBase ID: mmu-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: Chr. 18 - 65248861 - 65248926 [+] on Build GRCm38 |
| Stem-loop ID | rno-mir-122 |
| Stem-loop Accession # | MI0000891 |
| Stem-loop Sequence | CCUUAGCAGAGCUCUGGAGUGUGACAAUGGUGUUUGUGUCCAAAACAUCAAACGCCAUCAUCACACUAAACAGCUACUGCUAGGC |
| Chromosome Location | Chr. 18 - 60755285 - 60755369 [+] on Build Rnor_6.0 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000827
miRBase ID: rno-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: Chr. 18 - 60755285 - 60755369 [+] on Build Rnor_6.0 |
| Stem-loop ID | ami-mir-122 |
| Stem-loop Accession # | MI0029716 |
| Stem-loop Sequence | UACUAUCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAUCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038123
miRBase ID: ami-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | aca-mir-122 |
| Stem-loop Accession # | MI0018719 |
| Stem-loop Sequence | UCCUGCUGGAGCUGUGGAGUGUGACAAUGGUGUUUGUAUCCAAUCCGUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0021723
miRBase ID: aca-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | abu-mir-122 |
| Stem-loop Accession # | MI0033823 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUUUCCUCUUCGUCAAACGCCAUUAUCACACUAAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042117
miRBase ID: abu-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | bta-mir-122 |
| Stem-loop Accession # | MI0005063 |
| Stem-loop Sequence | CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003849
miRBase ID: bta-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | cja-mir-122 |
| Stem-loop Accession # | MI0032167 |
| Stem-loop Sequence | CUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039630
miRBase ID: cja-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | cpi-mir-122 |
| Stem-loop Accession # | MI0029454 |
| Stem-loop Sequence | CUGCCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAUCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0037724
miRBase ID: cpi-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | cli-mir-122 |
| Stem-loop Accession # | MI0029953 |
| Stem-loop Sequence | CUGCCCGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCUGGCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGGUAGACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038491
miRBase ID: cli-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | dno-mir-122 |
| Stem-loop Accession # | MI0038963 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUGUCCACAAUAUCAAACGCCAUUAUCACACUAAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047610
miRBase ID: dno-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | cfa-mir-122 |
| Stem-loop Accession # | MI0008015 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006619
miRBase ID: cfa-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | fru-mir-122 |
| Stem-loop Accession # | MI0003315 |
| Stem-loop Sequence | AGCUAUGGAGUGUGACAAUGGUGUUUGUUUCCUUUCUUUCAAACGCCAUUAUCACACUAAAUAGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002991
miRBase ID: fru-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | gmo-mir-122 |
| Stem-loop Accession # | MI0036011 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUUGUCUGAGCUUUCAAACGCCAUUAUCACACUAAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044063
miRBase ID: gmo-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | chi-mir-122 |
| Stem-loop Accession # | MI0030600 |
| Stem-loop Sequence | CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGGCAAUCCUUCCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0035918
miRBase ID: chi-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | ggo-mir-122 |
| Stem-loop Accession # | MI0020726 |
| Stem-loop Sequence | GAGUUUCCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGGCAAUCCUUCCCUCGAUAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0024179
miRBase ID: ggo-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | cpo-mir-122 |
| Stem-loop Accession # | MI0038621 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046961
miRBase ID: cpo-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | eca-mir-122 |
| Stem-loop Accession # | MI0012747 |
| Stem-loop Sequence | AGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUAGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0012995
miRBase ID: eca-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | mze-mir-122 |
| Stem-loop Accession # | MI0034121 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUUUCCUCUUCGUCAAACGCCAUUAUCACACUAAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042349
miRBase ID: mze-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | nbr-mir-122 |
| Stem-loop Accession # | MI0034406 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUUUCCUCUUCGUCAAACGCCAUUAUCACACUAAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042551
miRBase ID: nbr-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | oni-mir-122 |
| Stem-loop Accession # | MI0034648 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUUUCCUCUCCGUCAAACGCCAUUAUCACACUAAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042727
miRBase ID: oni-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | oan-mir-122 |
| Stem-loop Accession # | MI0006679 |
| Stem-loop Sequence | AACCUUGGAUUGUUUGCUACCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAGUCUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGUAGGGCCAUCCGUCUCUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006825
miRBase ID: oan-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | ptr-mir-122 |
| Stem-loop Accession # | MI0008431 |
| Stem-loop Sequence | CUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAAUUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0007963
miRBase ID: ptr-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | pol-mir-122 |
| Stem-loop Accession # | MI0022197 |
| Stem-loop Sequence | CUGUGGAGUGUGACAAUGGUGUUUGUGUCCUGUCUAUCAAACGCCAUUAUCACACUAUAUAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0025428
miRBase ID: pol-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | ppy-mir-122 |
| Stem-loop Accession # | MI0014815 |
| Stem-loop Sequence | CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUGCUAGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0015746
miRBase ID: ppy-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | pal-mir-122 |
| Stem-loop Accession # | MI0032322 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039776
miRBase ID: pal-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | pny-mir-122 |
| Stem-loop Accession # | MI0034859 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUUUCCUUUUCGUCAAACGCCAUUAUCACACUAAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042894
miRBase ID: pny-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | pbv-mir-122 |
| Stem-loop Accession # | MI0030210 |
| Stem-loop Sequence | UUCUGCUGGAGCUUUGGAGUGUGACAAUGGUGUUUGUAUCCAAUCUCUCAAACGCCAUUAUCACACUAAAUAGCUACUGUUAGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038909
miRBase ID: pbv-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | ocu-mir-122 |
| Stem-loop Accession # | MI0039275 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUGUCCAAACUAUCAAACGCCAUUAUCACACUAAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048201
miRBase ID: ocu-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | mml-mir-122a |
| Stem-loop Accession # | MI0007613 |
| Stem-loop Sequence | CCUUAGCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCUAAACUAUCAAACGCCAUUAUCACACUAAAUAGCUACUACUAGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0006180
miRBase ID: mml-miR-122a-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-122-1 |
| Stem-loop Accession # | MI0026450 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUGUCCUCUGCUGUCAAACGCCAUUAUUACACUAUAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032301
miRBase ID: ssa-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | ssa-mir-122-2 |
| Stem-loop Accession # | MI0026451 |
| Stem-loop Sequence | UGGAGUGUGACAAUGGUGUUUGUGUUCUCUGCAAUCAAACGCCAUUAUCACACUAAAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0032301
miRBase ID: ssa-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | tgu-mir-122 |
| Stem-loop Accession # | MI0013742 |
| Stem-loop Sequence | CCCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCAUCUAUCAAACGCCAUUAUCACACUAAAUAGUUCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0014529
miRBase ID: tgu-miR-122-5p Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | tni-mir-122 |
| Stem-loop Accession # | MI0003316 |
| Stem-loop Sequence | AGCUGUGGAGUGUGACAAUGGUGUUUGUGUCCUUUCUUUCAAACGCCAUUAUCACACUAAAUAGCU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002992
miRBase ID: tni-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
| Stem-loop ID | dre-mir-122 |
| Stem-loop Accession # | MI0001965 |
| Stem-loop Sequence | UCCAGAGCUGUGGAGUGUGACAAUGGUGUUUGUAUCAUCUGUCGUCAAACGCCAUUAUCACACUAAAUAGCCACGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0001818
miRBase ID: dre-miR-122 Mature miRNA Sequence: UGGAGUGUGACAAUGGUGUUUG Chromosome Location: NA |
More Information
Related Products
miRBase Alias:
Human: hsa-miR-122 (v17)
Mouse: mmu-miR-122 (v17)
Rat: rno-miR-122 (v18)
Aeromonas caviae Ae398: aca-miR-122 (v18)
Macaca mulatta: mml-miR-122a (v19)
Ochrobactrum anthropi: oan-miR-122 (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-122 (v19)
Mouse: mmu-miR-122 (v17)
Rat: rno-miR-122 (v18)
Aeromonas caviae Ae398: aca-miR-122 (v18)
Macaca mulatta: mml-miR-122a (v19)
Ochrobactrum anthropi: oan-miR-122 (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-122 (v19)

Genomic Map