Assay Details
Gene Family ID: | MIPF0000224, mir-287 |
Mature miRNA Sequence: | UGUGUUGAAAAUCGUUUGCAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-287-3p | MIMAT0000360 |
| Drosophila ananassae | dan-miR-287 | MIMAT0008424 |
| Drosophila erecta | der-miR-287 | MIMAT0008483 |
| Drosophila grimshawi | dgr-miR-287 | MIMAT0008563 |
| Drosophila mojavensis | dmo-miR-287 | MIMAT0008659 |
| Drosophila persimilis | dpe-miR-287 | MIMAT0008757 |
| View More Rows | ||
| Drosophila pseudoobscura | dps-miR-287 | MIMAT0001251 |
| Drosophila sechellia | dse-miR-287 | MIMAT0008770 |
| Drosophila simulans | dsi-miR-287 | MIMAT0008899 |
| Drosophila virilis | dvi-miR-287 | MIMAT0008930 |
| Drosophila willistoni | dwi-miR-287 | MIMAT0009003 |
Stem-loop Details
Drosophila melanogaster
Drosophila ananassae
Drosophila erecta
Drosophila grimshawi
Drosophila mojavensis
Drosophila persimilis
Drosophila pseudoobscura
Drosophila sechellia
Drosophila simulans
Drosophila virilis
Drosophila willistoni
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-287 (v16)

Genomic Map