Assay Details
Gene Family ID: | MIPF0000643, mir-658 |
Mature miRNA Sequence: | GGCGGAGGGAAGUAGGUCCGUUGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-658 | MIMAT0003336 |
| Pongo pygmaeus | ppy-miR-658 | MIMAT0016091 |
Stem-loop Details
Human
Pongo pygmaeus

Genomic Map