Assay Details
Gene Family ID: | MIPF0000001, mir-17 |
Mature miRNA Sequence: | UGCCCUAAAUGCCCCUUCUGGC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-18b-3p | MIMAT0004751 |
| Guinea pig | cpo-miR-18b-3p | MIMAT0046862 |
| Rabbit | ocu-miR-18b-3p | MIMAT0048114 |
Stem-loop Details
Human
Guinea pig
Rabbit

Genomic Map