Assay Details
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | UACUUGGAAAGGCAUCAGUUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-890 | MIMAT0004912 |
Pan troglodytes | ptr-miR-890 | MIMAT0008332 |
Pongo pygmaeus | ppy-miR-890 | MIMAT0016125 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
