Assay Details
Gene Family ID: | MIPF0000564, mir-1421 |
Mature miRNA Sequence: | AGGUGGUCACAUGAGUUGCAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1421n-5p | MIMAT0007074 |
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-1421n (v19)

Genomic Map