Assay Details
Mature miRNA Sequence: | AGAGCGAAGUGGGUGGGAAUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-1676-3p | MIMAT0007561 |
Stem-loop Details
Chicken
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | gga-miR-1676-3p |
|---|---|
| miRBase Accession Number: | MI0007410 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGAGCGAAGUGGGUGGGAAUGU |
| Species: | Chicken |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | AGAGCGAAGUGGGUGGGAAUGU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Chicken | gga-miR-1676-3p | MIMAT0007561 |