Assay Details
Mature miRNA Sequence: | UGAAAACUCAUUUUGCAUGCAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ornithorhynchus anatinus | oan-miR-1339-3p | MIMAT0006862 |
Stem-loop Details
Ornithorhynchus anatinus
More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-1339* (v18)

Genomic Map