Assay Details
Gene Family ID: | MIPF0000775, mir-1434 |
Mature miRNA Sequence: | GAAGAAAUCUAAGGUCUGAGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-1434-3p | MIMAT0012040 |
Stem-loop Details
Bovine

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bta-miR-1434-3p |
---|---|
miRBase Accession Number: | MI0011514 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | GAAGAAAUCUAAGGUCUGAGG |
Species: | Bovine |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000775, mir-1434 |
Mature miRNA Sequence: | GAAGAAAUCUAAGGUCUGAGG |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bovine | bta-miR-1434-3p | MIMAT0012040 |