Assay Details
Gene Family ID: | MIPF0000745, MIR2118 |
Mature miRNA Sequence: | UUCCUGAUGCCUCCCAUGCCUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Aegilops tauschii | ata-miR2118c-3p | MIMAT0037114 |
| Aegilops tauschii | ata-miR2118d-3p | MIMAT0037258 |
| Rice | osa-miR2118d | MIMAT0011743 |
| Sorghum bicolor | sbi-miR2118-3p | MIMAT0026427 |
| Zea mays | zma-miR2118d | MIMAT0011763 |
Stem-loop Details
Aegilops tauschii
Rice
Sorghum bicolor
Zea mays

Genomic Map