Assay Details
Gene Family ID: | MIPF0000812, mir-2188 |
Mature miRNA Sequence: | CUGUGUGAGGUUAGACCUAUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Zebrafish | dre-miR-2188-3p | MIMAT0011310 |
Stem-loop Details
Zebrafish
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dre-miR-2188-3p |
|---|---|
| miRBase Accession Number: | MI0010850 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUGUGUGAGGUUAGACCUAUC |
| Species: | Zebrafish |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000812, mir-2188 |
Mature miRNA Sequence: | CUGUGUGAGGUUAGACCUAUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Zebrafish | dre-miR-2188-3p | MIMAT0011310 |