Assay Details
Mature miRNA Sequence: | AUCAAGAUCGUUAGUAAGUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Schmidtea mediterranea | sme-miR-2203-3p | MIMAT0011420 |
Stem-loop Details
Schmidtea mediterranea
More Information
Related Products
miRBase Alias:
Schmidtea mediterranea: sme-miR-2203 (v18)

Genomic Map