Assay Details
Gene Family ID: | MIPF0000817, mir-2558 |
Mature miRNA Sequence: | CAGAUAUUUCUGUACCUACAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila pseudoobscura | dps-miR-2558-3p | MIMAT0012379 |
Stem-loop Details
Drosophila pseudoobscura
More Information
Related Products
miRBase Alias:
Derxomyces pseudoschimicola: dps-miR-2558* (v19)

Genomic Map