Assay Details
Gene Family ID: | MIPF0000254, mir-316 |
Mature miRNA Sequence: | UGUCUUUUUCCGCUUACUGCCG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Aedes aegypti | aae-miR-316 | MIMAT0014317 |
| Culex quinquefasciatus | cqu-miR-316 | MIMAT0014442 |
Stem-loop Details
Aedes aegypti
Culex quinquefasciatus

Genomic Map