Assay Details
Mature miRNA Sequence: | AAAAUCAAAUUGGGUUCUUUAUUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila pseudoobscura | dps-miR-2551-5p | MIMAT0012362 |
Stem-loop Details
Drosophila pseudoobscura
More Information
Related Products
miRBase Alias:
Derxomyces pseudoschimicola: dps-miR-2551* (v18)

Genomic Map