Assay Details
Gene Family ID: | MIPF0000293, mir-67 |
Mature miRNA Sequence: | UCACAACCUUUUUGAGUGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Nasonia vitripennis | nvi-miR-307 | MIMAT0015694 |
Stem-loop Details
Nasonia vitripennis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | nvi-miR-307 |
|---|---|
| miRBase Accession Number: | MI0014755 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCACAACCUUUUUGAGUGAG |
| Species: | Nasonia vitripennis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000293, mir-67 |
Mature miRNA Sequence: | UCACAACCUUUUUGAGUGAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Nasonia vitripennis | nvi-miR-307 | MIMAT0015694 |