Assay Details
Gene Family ID: | MIPF0000978, mir-2831 |
Mature miRNA Sequence: | UACAAACAAAGCAUUUGUUUGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2831 | MIMAT0013744 |
Stem-loop Details
Bombyx mori

Please enter the information below and press OK to send your cart to Core Services for purchase.
Assay Name: | bmo-miR-2831 |
---|---|
miRBase Accession Number: | MI0012428 |
miRBase Version: | v22.1 |
Mature miRNA Sequence: | UACAAACAAAGCAUUUGUUUGA |
Species: | Bombyx mori |
Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000978, mir-2831 |
Mature miRNA Sequence: | UACAAACAAAGCAUUUGUUUGA |
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Bombyx mori | bmo-miR-2831 | MIMAT0013744 |