Assay Details
Mature miRNA Sequence: | CGGCACCUAAUUGAAAUGCCCGCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-2495-3p | MIMAT0012205 |
Stem-loop Details
Drosophila melanogaster
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-2495* (v16)

Genomic Map