Assay Details
Gene Family ID: | MIPF0000006, mir-15 |
Mature miRNA Sequence: | CCAAUAUUGGCUGUGCUGCUCCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-195-3p | MIMAT0017149 |
| Dasypus novemcinctus | dno-miR-195-3p | MIMAT0047728 |
| Rabbit | ocu-miR-195-3p | MIMAT0048310 |
| Rhesus monkey | mml-miR-195-3p | MIMAT0026838 |
Stem-loop Details
Rat
Dasypus novemcinctus
Rabbit
Rhesus monkey

Genomic Map