Assay Details
Gene Family ID: | MIPF0001700, mir-2068 |
Mature miRNA Sequence: | GGACAUGUGUGCAUAGGGAAGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma floridae | bfl-miR-2068-3p | MIMAT0019170 |
Stem-loop Details
Branchiostoma floridae
More Information
Related Products
miRBase Alias:
Branchiostoma floridae: bfl-miR-2068* (v18)

Genomic Map