Assay Details
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | CGGACGGUAUAUGGGUUAAUAUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-311-5p | MIMAT0020837 |
Stem-loop Details
Drosophila melanogaster

Genomic Map