Assay Details
Mature miRNA Sequence: | UUUGUACUUUAUCAGGUGCUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Daphnia pulex | dpu-miR-305 | MIMAT0018924 |
Stem-loop Details
Daphnia pulex
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dpu-miR-305 |
|---|---|
| miRBase Accession Number: | MI0016448 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUGUACUUUAUCAGGUGCUCU |
| Species: | Daphnia pulex |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UUUGUACUUUAUCAGGUGCUCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Daphnia pulex | dpu-miR-305 | MIMAT0018924 |