Assay Details
Gene Family ID: | MIPF0000254, mir-316 |
Mature miRNA Sequence: | ACAGGAAAGGGAAAAAGGCGUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-316-3p | MIMAT0020842 |
| Drosophila virilis | dvi-miR-316-3p | MIMAT0032071 |
Stem-loop Details
Drosophila melanogaster
Drosophila virilis

Genomic Map