Assay Details
Gene Family ID: | MIPF0000134, mir-460 |
Mature miRNA Sequence: | ACAGCGCAUGCAAUGUGGAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-460b-3p | MIMAT0021973 |
Stem-loop Details
Anolis carolinensis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | aca-miR-460b-3p |
|---|---|
| miRBase Accession Number: | MI0018876 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | ACAGCGCAUGCAAUGUGGAC |
| Species: | Anolis carolinensis |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000134, mir-460 |
Mature miRNA Sequence: | ACAGCGCAUGCAAUGUGGAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-460b-3p | MIMAT0021973 |