Assay Details
Mature miRNA Sequence: | GAGCUGCCCUAUGAAGGACUGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ascaris suum | asu-miR-1822-3p | MIMAT0021536 |
Stem-loop Details
Ascaris suum
More Information
Related Products
miRBase Alias:
Atlantibacter subterranea: asu-miR-1822 (v18)

Genomic Map