Assay Details
Gene Family ID: | MIPF0000254, mir-316 |
Mature miRNA Sequence: | UGUCUUUUUCCGCUUUGCUGCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Manduca sexta | mse-miR-316 | MIMAT0024434 |
Stem-loop Details
Manduca sexta
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mse-miR-316 |
|---|---|
| miRBase Accession Number: | MI0020983 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUCUUUUUCCGCUUUGCUGCU |
| Species: | Manduca sexta |
| Product Type: | mirVana® miRNA mimic |
Gene Family ID: | MIPF0000254, mir-316 |
Mature miRNA Sequence: | UGUCUUUUUCCGCUUUGCUGCU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Manduca sexta | mse-miR-316 | MIMAT0024434 |