Assay Details
Mature miRNA Sequence: | UAAUCUGCAUCCUGAGGUUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Citrus sinensis | csi-miR2111-5p | MIMAT0049002 |
| Populus trichocarpa | ptc-miR2111a | MIMAT0025270 |
| Populus trichocarpa | ptc-miR2111b | MIMAT0025272 |
Stem-loop Details
Citrus sinensis
Populus trichocarpa

Genomic Map