Assay Details
Gene Family ID: | MIPF0000110, mir-329 |
Mature miRNA Sequence: | AACACACCUGGUUAACCUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Goat | chi-miR-329b-3p | MIMAT0036142 |
| Gorilla gorilla | ggo-miR-329b | MIMAT0024178 |
Stem-loop Details
Goat
Gorilla gorilla

Genomic Map