Assay Details
Mature miRNA Sequence: | UAGGCACCCUCGGUGAUACUUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila pseudoobscura | dps-miR-2512 | MIMAT0012297 |
Stem-loop Details
Drosophila pseudoobscura
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | dps-miR-2512 |
|---|---|
| miRBase Accession Number: | MI0011639 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGGCACCCUCGGUGAUACUUG |
| Species: | Drosophila pseudoobscura |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UAGGCACCCUCGGUGAUACUUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila pseudoobscura | dps-miR-2512 | MIMAT0012297 |